4-[(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)methyl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[(1,3-Dihydro-1-oxo-2H-isoindol-2-yl)methyl]benzoic acid

043752-5G
101862-394EA 602.6 USD
101862-394
4-[(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)methyl]benzenecarboxylic acid ≥95%
4-[(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)methyl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₃NO₃
MW: 267.29 g/mol
Melting Pt: 228…230 °C
MDL Number: MFCD06200936
CAS Number: 503039-50-7

Order Now

Specification Test Results

Melting point 228-230°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR