2-[5-Methyl-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(5-Methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)acetic acidThymin-1-ylacetic acid

043751-25G 043751-5G
101862-388EA 120 USD
101862-388 102833-122
2-[5-Methyl-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid ≥97%
2-[5-Methyl-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid

Hazard: Irritant

Formula: C₇H₈N₂O₄
MW: 184.15 g/mol
Melting Pt: 271…275 °C
MDL Number: MFCD00274250
CAS Number: 20924-05-4

Order Now

Specification Test Results

Melting point 271-275°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR