Methyl-4'-nitro-3'-(trifluoromethyl)[1,1'-biphenyl]-4-carboxylate ≥95%

Supplier: Matrix Scientific

043745-500MG
101862-362EA 278.46 USD
101862-362
Methyl-4'-nitro-3'-(trifluoromethyl)[1,1'-biphenyl]-4-carboxylate ≥95%
Methyl-4'-nitro-3'-(trifluoromethyl)[1,1'-biphenyl]-4-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₀F₃NO₄
Melting Pt: 143…145 °C
MDL Number: MFCD07698580
CAS Number: 886361-47-3

Order Now

Specification Test Results

Melting point 143-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR