2-Morpholinonicotinaldehyde ≥95%

Supplier: Matrix Scientific

043743-500MG 043743-1G
101862-352EA 747.69 USD
101862-352 101862-354
2-Morpholinonicotinaldehyde ≥95%
2-Morpholinonicotinaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MW: 192.22 g/mol
Melting Pt: 51…53 °C
MDL Number: MFCD03086195
CAS Number: 465514-09-4

Order Now

Specification Test Results

Melting point 51-53°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR