DL-β-(4-Benzyloxyphenyl)alaninol ≥95%
Supplier: Matrix Scientific
043719-500MG
043719-1G
101862-220EA
467.69
USD
101862-220
101862-218
DL-β-(4-Benzyloxyphenyl)alaninol ≥95%
DL-β-(4-Benzyloxyphenyl)alaninol
Hazard: Irritant
Formula:
C₁₆H₁₉NO₂ Melting Pt: 106…108 °C |
MDL Number:
MFCD04124850 CAS Number: 182057-85-8 |
Specification Test Results
Melting point | 106-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...