2-Amino-1-[4-(methylsulfonyl)phenyl]-1-ethanone hydrochloride ≥95%

Supplier: Matrix Scientific

043711-500MG
101862-182EA 921.54 USD
101862-182
2-Amino-1-[4-(methylsulfonyl)phenyl]-1-ethanone hydrochloride ≥95%
2-Amino-1-[4-(methylsulfonyl)phenyl]-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClNO₃S
Melting Pt: 300 °C
MDL Number: MFCD05663787
CAS Number: 102871-96-5

Order Now

Specification Test Results

Melting point 300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR