2-Ethyl-5-nitrobenzoxazole ≥95%

Supplier: Matrix Scientific

043702-500MG 043702-1G 043702-5G
101862-142EA 532.31 USD
101862-142 101862-140 101862-138
2-Ethyl-5-nitrobenzoxazole ≥95%
2-Ethyl-5-nitrobenzoxazole

Hazard: Irritant

Formula: C₉H₈N₂O₃
Melting Pt: 87…88 °C
CAS Number: 204771-74-4

Order Now

Specification Test Results

Melting point 87-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR