2-Ethyl-6-nitrobenzoxazole ≥95%
Supplier: Matrix Scientific
043699-500MG
043699-5G
043699-1G
101862-120EA
181.54
USD
101862-120
101862-122
101862-124
2-Ethyl-6-nitrobenzoxazole ≥95%
2-Ethyl-6-nitrobenzoxazole
Hazard: Irritant
Formula:
C₉H₈N₂O₃ Melting Pt: 83…85 °C |
CAS Number:
13243-39-5 |
Specification Test Results
Melting point | 83-85°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...