6-Nitropyrrolo[1,2-a]quinoline ≥95%

Supplier: Matrix Scientific

043698-1G 043698-500MG
101862-118EA 467.69 USD
101862-118 101862-116
6-Nitropyrrolo[1,2-a]quinoline ≥95%
6-Nitropyrrolo[1,2-a]quinoline

Hazard: Irritant

Formula: C₁₂H₈N₂O₂
Melting Pt: 140 °C
MDL Number: MFCD03791182
CAS Number: 52414-58-1

Order Now

Specification Test Results

Melting point 140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR