Methyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(2-amino-5-methylthiazol-4-yl)acetate

043694-1G 043694-5G
101862-100EA 153.85 USD
101862-100 101862-102
Methyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate ≥95%
Methyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Melting Pt: 111…113 °C
MDL Number: MFCD00662517
CAS Number: 259654-73-4

Order Now

Specification Test Results

Melting point 111-113°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR