5-Methyl-7-phenyl-6,7-dihydro-1H-1,4-diazepine-2,3-dicarbonitrile ≥95%
Supplier: Matrix Scientific
043690-500MG
043690-5G
043690-1G
101862-076EA
232.31
USD
101862-076
101862-078
101862-074
5-Methyl-7-phenyl-6,7-dihydro-1H-1,4-diazepine-2,3-dicarbonitrile ≥95%
5-Methyl-7-phenyl-6,7-dihydro-1H-1,4-diazepine-2,3-dicarbonitrile
Hazard: Irritant
Formula:
C₁₄H₁₂N₄ Melting Pt: 202…204 °C |
MDL Number:
MFCD03791179 CAS Number: 51802-61-0 |
Specification Test Results
Melting point | 202-204°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...