N-(7-Chloro-2,3-dihydro-1,4-benzodioxin-6-yl)acetamide ≥95%

Supplier: Matrix Scientific

043689-5G 043689-1G 043689-500MG
101862-068EA 181.54 USD
101862-068 101862-072 101862-070
N-(7-Chloro-2,3-dihydro-1,4-benzodioxin-6-yl)acetamide ≥95%
N-(7-Chloro-2,3-dihydro-1,4-benzodioxin-6-yl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₀ClNO₃
MW: 227.65 g/mol
Melting Pt: 173…175 °C
MDL Number: MFCD03791178
CAS Number: 855423-34-6

Order Now

Specification Test Results

Melting point 173-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR