4-(4-Chloroanilino)-2H-chromen-2-one ≥95%

Supplier: Matrix Scientific

043686-5G 043686-500MG 043686-1G
101862-062EA 532.31 USD
101862-062 101862-060 101862-058
4-(4-Chloroanilino)-2H-chromen-2-one ≥95%
4-(4-Chloroanilino)-2H-chromen-2-one

Hazard: Irritant

Formula: C₁₅H₁₀ClNO₂
Melting Pt: 299…301 °C
MDL Number: MFCD02325293
CAS Number: 24526-89-4

Order Now

Specification Test Results

Melting point 299-301°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR