4-Chloro-2-oxo-2H-chromene-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

043679-5G 043679-500MG 043679-1G
101862-032EA 232.31 USD
101862-032 101862-034 101862-030
4-Chloro-2-oxo-2H-chromene-3-carbaldehyde ≥95%
4-Chloro-2-oxo-2H-chromene-3-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₅ClO₃
Melting Pt: 127…129 °C
MDL Number: MFCD00179896
CAS Number: 50329-91-4

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR