2-Piperidinoisonicotinonitrile ≥95%

Supplier: Matrix Scientific

043676-1G 043676-5G 043676-500MG
101862-012EA 278.46 USD
101862-012 101862-014 101862-016
2-Piperidinoisonicotinonitrile ≥95%
2-Piperidinoisonicotinonitrile

Hazard: Irritant

Formula: C₁₁H₁₃N₃
Melting Pt: 58…60 °C
MDL Number: MFCD06200992
CAS Number: 127680-89-1

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR