1-Benzyl-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Benzyl-6-oxo-1,6-dihydropyridine-3-carboxylic acid

043656-1G 043656-500MG 043656-5G
101861-908EA 147.2 USD
101861-908 101861-906 101861-910
1-Benzyl-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%
1-Benzyl-6-oxo-1,6-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₁NO₃
MW: 229.24 g/mol
Melting Pt: 205…207 °C
MDL Number: MFCD00101801
CAS Number: 4332-79-0

Order Now

Specification Test Results

Melting point 205-207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR