2-(9,10-Dioxo-9,10-dihydro-2-anthracenyl)acetic acid ≥95%

Supplier: Matrix Scientific

043639-1G 043639-500MG
101861-828EA 1400 USD
101861-828 101861-826
2-(9,10-Dioxo-9,10-dihydro-2-anthracenyl)acetic acid ≥95%
2-(9,10-Dioxo-9,10-dihydro-2-anthracenyl)acetic acid

Hazard: Irritant

Formula: C₁₆H₁₀O₄
Melting Pt: 240…242 °C
MDL Number: MFCD00429262
CAS Number: 76161-80-3

Order Now

Specification Test Results

Melting point 240-242°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR