2-(2-Formyl-6-methoxyphenoxy)acetic acid ≥95%
Supplier: Matrix Scientific
043625-500MG
043625-1G
101861-756EA
443.08
USD
101861-756
101861-754
2-(2-Formyl-6-methoxyphenoxy)acetic acid ≥95%
2-(2-Formyl-6-methoxyphenoxy)acetic acid
Hazard: Irritant
Formula:
C₁₀H₁₀O₅ Melting Pt: 115…118 °C |
MDL Number:
MFCD00127291 CAS Number: 40359-30-6 |
Specification Test Results
Melting point | 115-118°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...