2-[3-(2-Nitro-1-propenyl)phenoxy]pyrimidine ≥95%

Supplier: Matrix Scientific

043621-5G 043621-500MG 043621-1G
101861-740EA 806.15 USD
101861-740 101861-738 101861-736
2-[3-(2-Nitro-1-propenyl)phenoxy]pyrimidine ≥95%
2-[3-(2-Nitro-1-propenyl)phenoxy]pyrimidine

Hazard: Irritant

Formula: C₁₃H₁₁N₃O₃
Melting Pt: 81…83 °C
MDL Number: MFCD03791172
CAS Number: 855308-69-9

Order Now

Specification Test Results

Melting point 81-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR