Methyl-2-chloro-4-morpholinobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

043611-5G 043611-500MG 043611-1G
101861-696EA 806.15 USD
101861-696 101861-694 101861-692
Methyl-2-chloro-4-morpholinobenzenecarboxylate ≥95%
Methyl-2-chloro-4-morpholinobenzenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₄ClNO₃
MW: 255.7 g/mol
Melting Pt: 69…70 °C
MDL Number: MFCD03012873
CAS Number: 175153-39-6

Order Now

Specification Test Results

Melting point 69-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR