N-(4-Ethynylphenyl)benzenesulfonamide ≥95%
Supplier: Matrix Scientific
043607-1G
043607-500MG
043607-5G
101861-674EA
553.85
USD
101861-674
101861-678
101861-676
N-(4-Ethynylphenyl)benzenesulfonamide ≥95%
N-(4-Ethynylphenyl)benzenesulfonamide
Hazard: Irritant
Formula:
C₁₄H₁₁NO₂S Melting Pt: 180…182 °C |
MDL Number:
MFCD03012024 CAS Number: 383147-75-9 |
Specification Test Results
Melting point | 180-182°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...