5-Chloro-2-thiophenecarboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chlorothiophene-2-carboxamide

043603-500MG 043603-1G
101861-652EA 330.77 USD
101861-652 101861-654
5-Chloro-2-thiophenecarboxamide ≥95%
5-Chloro-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₅H₄ClNOS
MW: 161.61 g/mol
Melting Pt: 179…181 °C
MDL Number: MFCD01922226
CAS Number: 22353-82-8

Order Now

Specification Test Results

Melting point 179-181°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR