7-Amino-6,7-dihydro-5H-indeno[5,6-d][1,3]dioxol-5-one hydrochloride ≥95%
Supplier: Matrix Scientific
043592-1G
043592-500MG
101861-592EA
927.69
USD
101861-592
101861-590
7-Amino-6,7-dihydro-5H-indeno[5,6-d][1,3]dioxol-5-one hydrochloride ≥95%
7-Amino-6,7-dihydro-5H-indeno[5,6-d][1,3]dioxol-5-one hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₀ClNO₃ Melting Pt: 270…273 °C (dec.) |
MDL Number:
MFCD04035629 CAS Number: 148502-20-9 |
Specification Test Results
Melting point | 270-273°C(d |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...