2,2,2-Trifluoro-N-(6-oxo-5,6-dihydro-4H-cyclopenta[b]thiophen-4-yl)acetamide ≥95%

Supplier: Matrix Scientific

043591-500MG 043591-5G 043591-1G
101861-586EA 927.69 USD
101861-586 101861-584 101861-588
2,2,2-Trifluoro-N-(6-oxo-5,6-dihydro-4H-cyclopenta[b]thiophen-4-yl)acetamide ≥95%
2,2,2-Trifluoro-N-(6-oxo-5,6-dihydro-4H-cyclopenta[b]thiophen-4-yl)acetamide

Hazard: Irritant

Formula: C₉H₆F₃NO₂S
MW: 249.21 g/mol
Melting Pt: 130…132 °C
MDL Number: MFCD03012801
CAS Number: 108046-14-6

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR