1-Methyl-5-(4-methylphenoxy)-3-phenyl-1H-pyrazole-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
043590-5G
043590-1G
043590-500MG
101861-582EA
1600
USD
101861-582
101861-580
101861-578
1-Methyl-5-(4-methylphenoxy)-3-phenyl-1H-pyrazole-4-carbaldehyde ≥95%
1-Methyl-5-(4-methylphenoxy)-3-phenyl-1H-pyrazole-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₈H₁₆N₂O₂ Melting Pt: 90…92 °C |
MDL Number:
MFCD03001208 CAS Number: 400073-98-5 |
Specification Test Results
Melting point | 90-92°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...