[1-Methyl-5-(4-methylphenoxy)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
Supplier: Matrix Scientific
043589-500MG
043589-1G
043589-5G
101861-574EA
927.69
USD
101861-574
101861-572
101861-576
[1-Methyl-5-(4-methylphenoxy)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
[1-Methyl-5-(4-methylphenoxy)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Hazard: Irritant
Formula:
C₁₃H₁₃F₃N₂O₂ Melting Pt: 88…90 °C |
MDL Number:
MFCD03001207 CAS Number: 400073-97-4 |
Specification Test Results
Melting point | 88-90°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...