Ethyl-N-[(5-amino-1-phenyl-1H-pyrazol-4-yl)carbonyl]ethanehydrazonoate ≥95%

Supplier: Matrix Scientific

043585-5G 043585-500MG 043585-1G
101861-552EA 806.15 USD
101861-552 101861-550 101861-548
Ethyl-N-[(5-amino-1-phenyl-1H-pyrazol-4-yl)carbonyl]ethanehydrazonoate ≥95%
Ethyl-N-[(5-amino-1-phenyl-1H-pyrazol-4-yl)carbonyl]ethanehydrazonoate

Hazard: Irritant

Formula: C₁₄H₁₇N₅O₂
Melting Pt: 146…148 °C
MDL Number: MFCD03001206
CAS Number: 99347-10-1

Order Now

Specification Test Results

Melting point 146-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR