N-(2,3-Dihydro-1H-inden-5-yl)-2-(4-hydroxyphenyl)acetamide ≥95%
Supplier: Matrix Scientific
043584-500MG
043584-1G
101861-546EA
232.31
USD
101861-546
101861-544
N-(2,3-Dihydro-1H-inden-5-yl)-2-(4-hydroxyphenyl)acetamide ≥95%
N-(2,3-Dihydro-1H-inden-5-yl)-2-(4-hydroxyphenyl)acetamide
Hazard: Irritant
Formula:
C₁₇H₁₇NO₂ MW: 267.33 g/mol Melting Pt: 157…158 °C |
MDL Number:
MFCD03425729 CAS Number: 321853-28-5 |
Specification Test Results
Melting point | 157-158°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...