6-(5-Chloro-2-hydroxyphenyl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

043578-500MG 043578-1G
101861-520EA 556.92 USD
101861-520 101861-522
6-(5-Chloro-2-hydroxyphenyl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid ≥95%
6-(5-Chloro-2-hydroxyphenyl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₄
Melting Pt: 257…259 °C
MDL Number: MFCD05021850
CAS Number: 439095-04-2

Order Now

Specification Test Results

Melting point 257-259°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR