2-(4-Chlorobenzyl)-1,3-thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

043571-500MG 043571-5G 043571-1G
101861-498EA 806.15 USD
101861-498 101861-496 101861-494
2-(4-Chlorobenzyl)-1,3-thiazole-4-carboxylic acid ≥95%
2-(4-Chlorobenzyl)-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₈ClNO₂S
Melting Pt: 193…195 °C
MDL Number: MFCD02102514
CAS Number: 477872-93-8

Order Now

Specification Test Results

Melting point 193-195°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR