2,4-Difluoro-5-nitrophenyl propionate ≥95%
Supplier: Matrix Scientific
043564-500MG
043564-1G
043564-5G
101861-464EA
806.15
USD
101861-464
101861-462
101861-460
2,4-Difluoro-5-nitrophenyl propionate ≥95%
2,4-Difluoro-5-nitrophenyl propionate
Hazard: Irritant
Formula:
C₉H₇F₂NO₄ Melting Pt: 42 °C |
MDL Number:
MFCD08443941 CAS Number: 924868-80-4 |
Specification Test Results
Melting point | 42°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...