Ethyl-1-(3,4-dichlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
043558-1G
043558-500MG
043558-5G
101861-442EA
181.54
USD
101861-442
101861-446
101861-444
Ethyl-1-(3,4-dichlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-(3,4-dichlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₄Cl₂N₂O₂ Melting Pt: 81…84 °C |
MDL Number:
MFCD02083022 CAS Number: 477710-51-3 |
Specification Test Results
Melting point | 81-84°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...