1-(4-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
043557-1G
043557-500MG
101861-438EA
350.77
USD
101861-438
101861-436
1-(4-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
1-(4-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₁FN₂O₂ Melting Pt: 215…217 °C |
MDL Number:
MFCD02083021 CAS Number: 288251-63-8 |
Specification Test Results
Melting point | 215-217°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...