3-[3-(2,5-Dimethyl-1H-pyrrol-1-yl)-2-thienyl]-3-oxopropanenitrile ≥95%
Supplier: Matrix Scientific
043550-500MG
043550-1G
043550-5G
101861-398EA
467.69
USD
101861-398
101861-396
101861-400
3-[3-(2,5-Dimethyl-1H-pyrrol-1-yl)-2-thienyl]-3-oxopropanenitrile ≥95%
3-[3-(2,5-Dimethyl-1H-pyrrol-1-yl)-2-thienyl]-3-oxopropanenitrile
Hazard: Irritant
Formula:
C₁₃H₁₂N₂OS Melting Pt: 156.5…158 °C |
MDL Number:
MFCD02102411 CAS Number: 477871-28-6 |
Specification Test Results
Melting point | 156.5-158°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...