Methyl-3-(2,5-dimethyl-1H-pyrrol-1-yl)-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

043547-1G 043547-5G 043547-500MG
101861-382EA 806.15 USD
101861-382 101861-380 101861-378
Methyl-3-(2,5-dimethyl-1H-pyrrol-1-yl)-2-thiophenecarboxylate ≥95%
Methyl-3-(2,5-dimethyl-1H-pyrrol-1-yl)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₃NO₂S
Melting Pt: 142…144 °C
MDL Number: MFCD00053053
CAS Number: 175137-41-4

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR