2-[5-(2-Fluorophenyl)-6-methyl-4-oxo-2H-1,3-oxazin-3(4H)-yl]-2-methylpropanoic acid ≥95%
Supplier: Matrix Scientific
043535-1G
043535-500MG
101861-322EA
921.54
USD
101861-322
101861-324
2-[5-(2-Fluorophenyl)-6-methyl-4-oxo-2H-1,3-oxazin-3(4H)-yl]-2-methylpropanoic acid ≥95%
2-[5-(2-Fluorophenyl)-6-methyl-4-oxo-2H-1,3-oxazin-3(4H)-yl]-2-methylpropanoic acid
Hazard: Irritant
Formula:
C₁₅H₁₆FNO₄ Melting Pt: 193 °C |
MDL Number:
MFCD11553017 CAS Number: 170855-30-8 |
Specification Test Results
Melting point | 193°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...