3-[2-(4-Chloro-3-fluorophenoxy)phenyl]acrylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
trans-2-(4-Chloro-3-fluorophenoxy)cinnamic acid
043529-500MG
043529-1G
043529-5G
101861-288EA
927.69
USD
101861-288
101861-286
101861-290
3-[2-(4-Chloro-3-fluorophenoxy)phenyl]acrylic acid ≥95%
3-[2-(4-Chloro-3-fluorophenoxy)phenyl]acrylic acid
Hazard: Irritant
Formula:
C₁₅H₁₀ClFO₃ MW: 292.7 g/mol Melting Pt: 197…199 °C |
MDL Number:
MFCD02091010 CAS Number: 449778-67-0 |
Specification Test Results
Melting point | 197-199°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...