3-[2-(3-Chlorophenoxy)phenyl]acrylic acid ≥95%
Supplier: Matrix Scientific
043525-1G
043525-500MG
043525-5G
101861-266EA
1600
USD
101861-266
101861-264
101861-262
3-[2-(3-Chlorophenoxy)phenyl]acrylic acid ≥95%
3-[2-(3-Chlorophenoxy)phenyl]acrylic acid
Hazard: Irritant
Formula:
C₁₅H₁₁ClO₃ Melting Pt: 175…177 °C |
MDL Number:
MFCD02091007 CAS Number: 338393-65-0 |
Specification Test Results
Melting point | 175-177°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...