Methyl-4-[(1H-1,2,4-triazol-3-ylsulfanyl)methyl]phenyl ether ≥95%
Supplier: Matrix Scientific
043513-500MG
043513-5G
043513-1G
101861-198EA
1073.85
USD
101861-198
101861-196
101861-194
Methyl-4-[(1H-1,2,4-triazol-3-ylsulfanyl)methyl]phenyl ether ≥95%
Methyl-4-[(1H-1,2,4-triazol-3-ylsulfanyl)methyl]phenyl ether
Hazard: Irritant
Formula:
C₁₀H₁₁N₃OS Melting Pt: 85…87 °C |
MDL Number:
MFCD01211112 CAS Number: 17357-75-4 |
Specification Test Results
Melting point | 85-87°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...