4-Chloro-5-[(2-hydroxyethyl)(methyl)amino]-2-(4-methoxyphenyl)-3(2H)-pyridazinone ≥95%
Supplier: Matrix Scientific
043493-5G
043493-1G
043493-500MG
101861-088EA
467.69
USD
101861-088
101861-086
101861-090
4-Chloro-5-[(2-hydroxyethyl)(methyl)amino]-2-(4-methoxyphenyl)-3(2H)-pyridazinone ≥95%
4-Chloro-5-[(2-hydroxyethyl)(methyl)amino]-2-(4-methoxyphenyl)-3(2H)-pyridazinone
Hazard: Irritant
Formula:
C₁₄H₁₆ClN₃O₃ Melting Pt: 162…164 °C |
MDL Number:
MFCD00793824 CAS Number: 449778-65-8 |
Specification Test Results
Melting point | 162-164°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...