7-[1-(3-Chlorophenoxy)ethyl][1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%
Supplier: Matrix Scientific
043487-5G
043487-1G
043487-500MG
101861-054EA
2129.23
USD
101861-054
101861-052
101861-050
7-[1-(3-Chlorophenoxy)ethyl][1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%
7-[1-(3-Chlorophenoxy)ethyl][1,2,4]triazolo[1,5-a]pyrimidin-2-amine
Hazard: Irritant
Formula:
C₁₃H₁₂ClN₅O MW: 289.73 g/mol Melting Pt: 211…213 °C |
MDL Number:
MFCD00243648 CAS Number: 477865-76-2 |
Specification Test Results
Melting point | 211-213°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...