N-Acetylguanidine ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Carbamimidoylacetamide
, 1-Acetylguanidine
043484-25G
043484-5G
101861-036EA
80
USD
101861-036
101861-034
N-Acetylguanidine ≥95%
N-Acetylguanidine
Hazard: Irritant/Stored under Argon
Formula:
C₃H₇N₃O MW: 101.11 g/mol Melting Pt: 188…190 °C |
MDL Number:
MFCD00075615 CAS Number: 5699-40-1 |
Specification Test Results
Melting point | 188-190°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...