4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methyl}benzenol ≥95%
Supplier: Matrix Scientific
043473-1G
043473-5G
043473-500MG
101860-974EA
747.69
USD
101860-974
101860-976
101860-978
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methyl}benzenol ≥95%
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methyl}benzenol
Hazard: Irritant
Formula:
C₁₃H₉ClF₃NO MW: 287.67 g/mol Melting Pt: 135…137 °C |
MDL Number:
MFCD01316641 CAS Number: 321432-62-6 |
Specification Test Results
Melting point | 135-137°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...