1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
043465-5G
043465-500MG
043465-1G
101860-940EA
2129.23
USD
101860-940
101860-936
101860-938
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrole-2-carbaldehyde ≥95%
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrole-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₁H₆ClF₃N₂O MW: 274.63 g/mol Melting Pt: 60…63 °C |
MDL Number:
MFCD00231912 CAS Number: 321430-69-7 |
Specification Test Results
Melting point | 60-63°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...