1-[(Ammoniooxy)methyl]-2-methoxybenzene chloride ≥95%
Supplier: Matrix Scientific
Synonyms:
O-[(2-Methoxyphenyl)methyl]hydroxylamine hydrochloride
043461-1G
043461-500MG
043461-5G
101860-920EA
467.69
USD
101860-920
101860-922
101860-918
1-[(Ammoniooxy)methyl]-2-methoxybenzene chloride ≥95%
1-[(Ammoniooxy)methyl]-2-methoxybenzene chloride
Hazard: Irritant
Formula:
C₈H₁₂ClNO₂ Melting Pt: 112…116 °C |
MDL Number:
MFCD01114579 CAS Number: 317821-72-0 |
Specification Test Results
Melting point | 112-116°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...