1-[(Ammoniooxy)methyl]-2-(trifluoromethyl)benzene chloride ≥95%
Supplier: Matrix Scientific
Synonyms:
O-[2-(Trifluoromethyl)benzyl]hydroxylamine hydrochloride
043460-5G
043460-500MG
043460-1G
101860-912EA
615.38
USD
101860-912
101860-914
101860-916
1-[(Ammoniooxy)methyl]-2-(trifluoromethyl)benzene chloride ≥95%
1-[(Ammoniooxy)methyl]-2-(trifluoromethyl)benzene chloride
Hazard: Irritant
Formula:
C₈H₉ClF₃NO Melting Pt: 135…140 °C |
MDL Number:
MFCD01114578 CAS Number: 215599-92-1 |
Specification Test Results
Melting point | 135-140°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...