1-[(Ammoniooxy)methyl]-2-chloro-4-fluorobenzene chloride ≥95%
Supplier: Matrix Scientific
Synonyms:
O-(2-Chloro-4-fluorobenzyl)hydroxylamine hydrochloride
043455-5G
043455-1G
043455-500MG
101860-886EA
2129.23
USD
101860-886
101860-882
101860-884
1-[(Ammoniooxy)methyl]-2-chloro-4-fluorobenzene chloride ≥95%
1-[(Ammoniooxy)methyl]-2-chloro-4-fluorobenzene chloride
Hazard: Irritant
Formula:
C₇H₈Cl₂FNO Melting Pt: 175…180 °C |
MDL Number:
MFCD01114574 CAS Number: 317821-68-4 |
Specification Test Results
Melting point | 175-180°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...