2-[(3-Nitro-2-pyridinyl)sulfanyl]aniline ≥95%

Supplier: Matrix Scientific

043449-500MG 043449-1G 043449-5G
101860-852EA 67.69 USD
101860-852 101860-854 101860-856
2-[(3-Nitro-2-pyridinyl)sulfanyl]aniline ≥95%
2-[(3-Nitro-2-pyridinyl)sulfanyl]aniline

Hazard: Irritant

Formula: C₁₁H₉N₃O₂S
Melting Pt: 125.5…127.5 °C
MDL Number: MFCD01936009
CAS Number: 92316-06-8

Order Now

Specification Test Results

Melting point 125.5-127.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR