4-(3,4-Dimethoxyphenyl)-3-methyl-1H-pyrazole ≥95%

Supplier: Matrix Scientific

043444-500MG 043444-1G 043444-5G
101860-822EA 322 USD
101860-822 101860-824 101860-826
4-(3,4-Dimethoxyphenyl)-3-methyl-1H-pyrazole ≥95%
4-(3,4-Dimethoxyphenyl)-3-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂
Melting Pt: 114…116 °C
MDL Number: MFCD00112345
CAS Number: 321526-09-4

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR