2,4-Dioxo-1-piperidino-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%

Supplier: Matrix Scientific

043438-500MG 043438-1G
101860-794EA 1109.23 USD
101860-794 101860-796
2,4-Dioxo-1-piperidino-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%
2,4-Dioxo-1-piperidino-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile

Hazard: Irritant

Formula: C₁₀H₁₂N₄O₂
Melting Pt: 265…269 °C
MDL Number: MFCD00172932
CAS Number: 320420-05-1

Order Now

Specification Test Results

Melting point 265-269°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR